Merck
  • Decreased lipogenesis in white adipose tissue contributes to the resistance to high fat diet-induced obesity in phosphatidylethanolamine N-methyltransferase-deficient mice.
Decreased lipogenesis in white adipose tissue contributes to the resistance to high fat diet-induced obesity in phosphatidylethanolamine N-methyltransferase-deficient mice.
Biochimica et biophysica acta (2014-12-03)
Xia Gao, Jelske N van der Veen, Martin Hermansson, Marta Ordoñez, Antonio Gomez-Muñoz, Dennis E Vance, René L Jacobs
ZUSAMMENFASSUNG
Mice lacking phosphatidylethanolamine N-methyltransferase (PEMT, Pemt(-/-) mice) are resistant to high-fat diet (HFD)-induced obesity (DIO) but develop non-alcoholic steatohepatitis. PEMT expression is strongly induced during differentiation of 3T3-L1 adipocytes. Hence, we hypothesized that white adipose tissue (WAT) might be a key player in the protection against DIO in Pemt(-/-) mice. We fed Pemt(-/-) and Pemt(+/+) mice the HFD for 2 weeks, after which we examined adipocyte differentiation, adipogenesis and lipolysis in WAT. Pemt(-/-) mice gained less body weight, had reduced WAT mass and had smaller adipocytes than Pemt(+/+) mice. The protein levels of adipose differentiation markers FABP4, PPARγ and C/EBPβ were not altered by genotype, but acetyl-CoA carboxylase expression and activation was reduced in the Pemt(-/-) mice. The in vivo conversion of [¹⁴C]acetate to [¹⁴C]TG in WAT was also lower in Pemt(-/-) mice. The release of glycerol from WAT explants was comparable between Pemt(+/+) and Pemt(-/-) mice under basal condition and in the presence of isoproterenol, indicating unaffected lipolytic capacity. Furthermore, the amounts of leptin, cytokines and chemokines in WAT were not altered by genotype in mice fed the HFD for 2 weeks. However, after 10 weeks of HFD, WAT from Pemt(-/-) mice had dramatically lower leptin, inflammatory cytokines (IL-1 and TNF-α) and chemokines (MCP-1 and RANTES), and significantly higher anti-inflammatory cytokine IL-10 than Pemt(+/+) mice. Together, our data show that PEMT deficiency did not affect the capability for differentiation and lipolysis in WAT. Decreased lipogenesis in WAT may contribute to the resistance to DIO in Pemt(-/-) mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture