Merck
  • The impact of storage temperature on the morphology, viability, cell number and metabolism of cultured human conjunctival epithelium.
The impact of storage temperature on the morphology, viability, cell number and metabolism of cultured human conjunctival epithelium.
Current eye research (2014-04-23)
Jon R Eidet, Øygunn A Utheim, Rakibul Islam, Torstein Lyberg, Edvard B Messelt, Darlene A Dartt, Tor P Utheim
ZUSAMMENFASSUNG
To evaluate the effect of storage temperature on the morphology, viability, cell number and metabolism of cultured human conjunctival epithelial cells (HCjEs). Three-day cultured HCjEs were stored at nine different temperatures between 4 °C and 37 °C for four and seven days. Phenotype was assessed by immunofluorescence microscopy, morphology by scanning electron microscopy, viability and cell number by a microplate fluorometer and glucose metabolism by a blood gas analyzer. Cultured cells not subjected to storage expressed the conjunctival cytokeratins 7 and 19 and the proliferation marker proliferating cell nuclear antigen. Cell morphology was best maintained following four-day storage between 12 °C and 28 °C and following 12 °C storage after seven days. Assessed by propidium iodide uptake, the percentage of viable cells after four-day storage was maintained only between 12 °C and 28 °C, whereas it had decreased in all other groups (p < 0.05; n = 4). After seven days this percentage was maintained in the 12 °C group, but it had decreased in all other groups, compared to the control (p < 0.05; n = 4). The total number of cells remaining in the cultures after four-day storage, compared to the control, had declined in all groups (p < 0.05; n = 4), except 12 °C and 20 °C groups. Following seven days this number had decreased in all groups (p < 0.01; n = 4), except 12 °C storage. Four-day storage at 12 °C demonstrated superior preservation of the number of calcein-stained viable cells (p < 0.05) and the least accumulation of ethidium homodimer 1-stained dead cells (p < 0.001), compared to storage at 4 °C and 24 °C (n = 6). The total metabolism of glucose to lactate after four-day storage was higher in the 24 °C group compared to 4 °C and 12 °C groups, as well as the control (p < 0.001; n = 3). Storage at 12 °C appears optimal for preserving the morphology, viability and total cell number in stored HCjE cultures. The superior cell preservation at 12 °C may be related to temperature-associated effects on cell metabolism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Ethidiumhomodimer, suitable for fluorescence, ~90% (HPCE)
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
2-Phenylindol, technical grade, 95%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material