Merck
  • Characterization of Melanoplus sanguinipes oviposition stimulating protein expression and re-examination of its potential role in stimulating oviposition.
Characterization of Melanoplus sanguinipes oviposition stimulating protein expression and re-examination of its potential role in stimulating oviposition.
Journal of insect physiology (2015-01-27)
E A Sieminska, J A Andres, C D Todd, M A Erlandson
ZUSAMMENFASSUNG
Melanoplussanguinipes oviposition stimulating protein (MsOSP) was characterized and its role in stimulating oviposition in virgin females was examined. A 967nt MsOSP mRNA sequence with homology to previously characterized N-terminal amino acid sequence data for MsOSP was identified in a RNAseq library generated from an mRNA pool from the long hyaline tubule (LHT) of the male accessory gland complex. This transcript contained a predicted 729nt open reading frame encoding the 242aa putative MsOSP protein and had the second highest read abundance in the library. The MsOSP transcript was detected exclusively in the LHT tissue of adult males and its abundance increased with time until 7 days post-eclosion. Western blot analysis using an anti-MsOSP antibody showed high levels of MsOSP protein in the LHT luminal secretions of virgin males and to a lesser degree was associated with the aedeagus and ejaculatory duct. MsOSP was shown to be a major protein component of the spermatophore packet transferred from the male to female during copulation. However, only minor amounts of MsOSP could be detected in the female bursa, spermatheca and oviduct. Intrahemocoelic injection of LHT luminal protein into mature virgin females stimulated oviposition in ∼ 65% of females. A similar but non-significant trend was observed upon injection of purified recombinant MsOSP protein, and immunoprecipitation of LHT protein with anti-MsOSP antibody led to abrogation of oviposition stimulation upon injection of mature virgin females. Despite the demonstration of stimulation of oviposition upon intrahemocoelic injection of LHT-derived-MsOSP into mature virgin females, the potential mode of action of MsOSP in this process remains to be determined. MsOSP cannot be detected in the tissues other than the bursa, spermatheca and oviduct of female grasshoppers and relatively large quantities of MsOSP are required to stimulate oviposition upon injection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
1H-1,2,3-Triazol, 97%
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)