Merck
  • Improved biosynthesis of ethyl (S)-4-chloro-3-hydroxybutanoate by adding L-glutamine plus glycine instead of NAD+ in β-cyclodextrin-water system.
Improved biosynthesis of ethyl (S)-4-chloro-3-hydroxybutanoate by adding L-glutamine plus glycine instead of NAD+ in β-cyclodextrin-water system.
Bioresource technology (2015-02-16)
Yu-Cai He, Dan-Ping Zhang, Zhi-Cheng Tao, Yun Lu, Yun Ding, Feng Liu, Zheng-Zhong Zhu, Huan Rui, Gao-Wei Zheng, Xian Zhang
ZUSAMMENFASSUNG
To reduce dependence on the expensive cofactor and effectively biotransform ethyl 4-chloro-3-oxobutanoate, L-glutamine and glycine were found to enhance the content of intracellular NADH and the reductase activity. Adding the mixture of 200 mM of L-glutamine and 500 mM of glycine to the reaction media, a 1.67-fold of reductase activity was increased over the control without the addition of the two compounds. Moreover, β-cyclodextrin (0.4 mol β-cyclodextrin/mol ethyl 4-chloro-3-oxobutanoate) was also added into this reaction media, and the biocatalytic activity of the whole-cell biocatalyst of Escherichia coli CCZU-K14 was increased by 1.34-fold than that without β-cyclodextrin. In this β-cyclodextrin-water media containing L-glutamine (200 mM) plus glycine (500 mM), ethyl (S)-4-chloro-3-hydroxybutanoate (>99% ee) could be obtained from 3000 mM ethyl 4-chloro-3-oxobutanoate in the yield of 98.0% after 8h. All the positive features demonstrate the potential applicability of the bioprocess for the large-scale production of ethyl (S)-4-chloro-3-hydroxybutanoate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
SAFC
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycin, 99%, FCC
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method