Merck
The possible involvement of skin dryness on alterations of the dermal matrix.
Experimental dermatology (2014-09-23)
Mariko Yokota, Kenji Shimizu, Daiki Kyotani, Shoichi Yahagi, Satoru Hashimoto, Hitoshi Masaki
ZUSAMMENFASSUNG
Moisturization of the skin plays an important role in maintaining skin homeostasis. Although it is understood that skin dryness initiates the formation of fine wrinkles, there are few objective reports to support that understanding. The purpose of this study was to establish an in vitro dry epidermal model using reconstructed human epidermal equivalents (RHEEs) and to elucidate the relationship between skin dryness and alterations of the dermal matrix which is one of the causes for the formation of wrinkles. An in vitro dry epidermal model was prepared by loading a CaCl2 -filled ampoule on the surface of an RHEE. To evaluate whether the in vitro model reproduced the characteristics of in vivo skin dryness, histological studies and biological assays using a protein array were carried out. Histologically, a distinct fluorescence which originated from carbonylated protein was observed in the stratum corneum. In addition, conditioned medium from RHEEs cultured under dry conditions for 24 h revealed the secretion of several proteins, such as IL-1α, IL-1ra, IL-8, MMP-9, VEGF, M-CSF and IGFBP-2 and IGFBP-3, galectin-1, Cys-C, FGF-6, OPG, Glc and TSP4 compared with normal RHEEs. It has been reported that an increase in IL-1α and the accumulation of carbonylated proteins are observed in the intact stratum corneum in the low humidity winter season. Thus, the in vitro dry epidermal model expresses the features of in vivo skin dryness observed in the winter season. Furthermore, the conditioned medium from RHEEs cultured under dry conditions enhanced MMP-1 secretion by normal human dermal fibroblasts. Taken together, we propose the hypothesis that skin dryness contributes to alterations of the dermal matrix through the elevation of MMP-1 secretion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Eosin Y -Lösung, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y Dinatriumsalz, certified by the BSC
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Eosin Y Dinatriumsalz, ≥85 % by HPLC
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Fluoreszein-5-thiosemicarbazid, suitable for fluorescence, ~80% (HPCE)
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis