Merck
  • Microencapsulation of probiotic bacteria Lactobacillus plantarum 15HN using alginate-psyllium-fenugreek polymeric blends.
Microencapsulation of probiotic bacteria Lactobacillus plantarum 15HN using alginate-psyllium-fenugreek polymeric blends.
Journal of applied microbiology (2015-01-27)
B Haghshenas, N Abdullah, Y Nami, D Radiah, R Rosli, A Yari Khosroushahi
ZUSAMMENFASSUNG
Investigation on the use of herbal-based biopolymers for probiotic-Lactobacillus plantarum 15HN-encapsulation is presented. The objectives are to enhance its oral delivery, colonic release and survival rate of these probiotic cultures in gastrointestinal environment. Nine types of herbal-based polymers blend with different concentration of alginate alone or mixed with psyllium and fenugreek was used as candidate for encapsulation matrix by applying a simple extrusion method. All the blend formulations recorded high encapsulation efficiency at value >98%. The survival rate of viable probiotic cells under both low pH and high bile salt conditions was also high with value above 80% in 2% (w/v) alginate, alginate+psyllium (1·5 + 0·5%) blend and alginate+fenugreek (1·5 + 0·5%) blend as compared to other polymer formulations and nonencapsulated cells. Their release occurred after 2 h in colonic condition and sustained until the 12th hour incubation period. A value added prebiotic effect was observed in (1·5 + 0·5%) alginate-psyllium formulation. The high encapsulation efficiency, high viability of cell in low pH, high bile salt and the sustained release rates of probiotic cells in colonic condition during storage time was also observed for these herbal gel formulations. Herbal-based biopolymers offer added advantages of being prebiotic towards the enhancement of probiotic bacterial growth in the gastrointestinal environment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumphosphat, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Natriumphosphat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Natronlauge, 5.0 M
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Sigma-Aldrich
Natriumphosphat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%