Merck
  • Characterizations of plasticized polymeric film coatings for preparing multiple-unit floating drug delivery systems (muFDDSs) with controlled-release characteristics.
Characterizations of plasticized polymeric film coatings for preparing multiple-unit floating drug delivery systems (muFDDSs) with controlled-release characteristics.
PloS one (2014-06-27)
Sheng-Feng Hung, Chien-Ming Hsieh, Ying-Chen Chen, Yu-Chun Wang, Hsiu-O Ho, Ming-Thau Sheu
ZUSAMMENFASSUNG
Effervescent multiple-unit floating drug delivery systems (muFDDSs) consisting of drug (lorsartan)- and effervescent (sodium bicarbonate)-containing pellets were characterized in this study. The mechanical properties (stress and strain at rupture, Young's modulus, and toughness) of these plasticized polymeric films of acrylic (Eudragit RS, RL, and NE) and cellulosic materials (ethyl cellulose (EC), and Surelease) were examined by a dynamic mechanical analyzer. Results demonstrated that polymeric films prepared from Surelease and EC were brittle with less elongation compared to acrylic films. Eudragit NE films were very flexible in both the dry and wet states. Because plasticizer leached from polymeric films during exposure to the aqueous medium, plasticization of wet Eudragit RS and RL films with 15% triethyl citrate (TEC) or diethyl phthalate (DEP) resulted in less elongation. DEP might be the plasticizer of choice among the plasticizers examined in this study for Eudragit RL to provide muFDDSs with a short time for all pellets to float (TPF) and a longer period of floating. Eudragit RL and RS at a 1∶1 ratio plasticized with 15% DEP were optimally selected as the coating membrane for the floating system. Although the release of losartan from the pellets was still too fast as a result of losartan being freely soluble in water, muFDDSs coated with Eudragit RL and RS at a 1∶1 ratio might have potential use for the sustained release of water-insoluble or the un-ionized form of drugs from gastroretentive drug delivery systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Phthalsäuredibutylester, 99%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Diethylphthalat, 99.5%
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triethylcitrat, ≥98.0% (GC)
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, tested according to Ph. Eur.
Sigma-Aldrich
Triethylcitrat, ≥99%, FCC, FG
Supelco
Diethyl-Phthalat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethylphthalat, ≥99%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Supelco
Triethylcitrat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phthalsäuredibutylester, PESTANAL®, analytical standard
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Supelco
Diethylphthalat, PESTANAL®, analytical standard
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
DBP, 98% (HPLC)
Supelco
Phthalsäuredibutylester, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Phthalsäuredibutylester, European Pharmacopoeia (EP) Reference Standard
Diethylphthalat, European Pharmacopoeia (EP) Reference Standard