Merck
  • Oxidation potentials of N-modified derivatives of the analgesic flupirtine linked to potassium KV 7 channel opening activity but not hepatocyte toxicity.
Oxidation potentials of N-modified derivatives of the analgesic flupirtine linked to potassium KV 7 channel opening activity but not hepatocyte toxicity.
ChemMedChem (2014-11-14)
Christian J Lemmerhirt, Mirko Rombach, Anja Bodtke, Patrick J Bednarski, Andreas Link
ZUSAMMENFASSUNG
Openers of neuronal voltage-gated potassium channels (KV ) are of interest as therapeutic agents for treating pain (flupirtine) and epilepsy (retigabine). In an effort to better understand the mechanisms of action and toxicity of flupirtine, we synthesized nine novel analogues with varying redox behavior. Flupirtine can be oxidatively metabolized into azaquinone diimines; thus, the oxidation potentials of flupirtine and its analogues were measured by cyclic voltammetry. KV 7.2/3 (KCNQ2/3) opening activity was determined by an established assay with HEK293 cells overexpressing these channels. A link was found between the oxidation potentials of the compounds and their EC50 values for potassium channel opening activity. On the other hand, no correlation was observed between oxidation potentials and cytotoxicity in cultures of transgenic mouse hepatocytes (TAMH). These results support the idea that oxidative metabolites of flupirtine contribute to the mechanism of action, similar to what was recently proposed for acetaminophen (paracetamol), but not to hepatotoxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamid, ≥99.5% (HPLC)
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Supelco
Niacinamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamid, ≥98% (HPLC), powder
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
USP
Niacinamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Niacinamid, meets USP testing specifications
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Kaliumhydrid, in paraffin
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Supelco
Streptomycin -Lösung, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Nicotinamid, ≥98.5% (HPLC)
Nicotinamid, European Pharmacopoeia (EP) Reference Standard
Streptomycinsulfat, European Pharmacopoeia (EP) Reference Standard
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.