Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Sigma-Aldrich
Aluminiumchlorid, ReagentPlus®, 99%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Aluminiumchlorid, reagent grade, 98%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Methansulfonylchlorid, ≥99.7%
Sigma-Aldrich
Methylamin -Lösung, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Thionylchlorid, ReagentPlus®, ≥99%
Sigma-Aldrich
Methylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Methylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Aluminiumchlorid, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Aluminiumchlorid, 99.99% trace metals basis
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thionylchlorid, reagent grade, 97%
Supelco
Dimethylformamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminiumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Methylamin -Lösung, 2.0 M in methanol
Sigma-Aldrich
Aluminiumchlorid -Lösung, 1.0 M in nitrobenzene
Supelco
N,N-Dimethylformamid, analytical standard
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure, technical grade
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure
Sigma-Aldrich
Methansulfonylchlorid, purum, ≥98.0% (AT)