Merck
  • Daily consumption of banana marginally improves blood glucose and lipid profile in hypercholesterolemic subjects and increases serum adiponectin in type 2 diabetic patients.
Daily consumption of banana marginally improves blood glucose and lipid profile in hypercholesterolemic subjects and increases serum adiponectin in type 2 diabetic patients.
Indian journal of experimental biology (2015-02-06)
Ratchada Cressey, Warunee Kumsaiyai, Ampika Mangklabruks
ZUSAMMENFASSUNG
In this study, we explored the effects of consumption of banana in thirty hypercholesterolemic and fifteen type 2 diabetic subjects. They were given a daily dose of 250 or 500 grams of banana for breakfast for 12 weeks. Fasting serum lipid, glucose and insulin levels were measured initially as well as every 4 weeks. Daily consumption of banana significantly lowered fasting blood glucose (from 99 ± 7.7 to 92 ± 6.9 and 102 ± 7.3 to 92 ± 5.7 mg x dL(-1) (p < 0.05) after consuming banana 250 or 500 g/day for 4 wk, respectively) and LDL-cholesterol/HDL-cholesterol ratio (from 2.7 ± 0.98 to 2.4 ± 0.85 and 2.8 ± 0.95 to 2.5 ± 0.79, p < 0.005) in hypercholesterolemic volunteers. Analysis of blood glycemic response after eating banana showed significantly lower 2 h-postprandial glucose level compared to baseline in hypercholesterolemic volunteers given a dose of 250 g/day. The changes of blood glucose and lipid profile in diabetic patients were not statistically significant, but for plasma levels of adiponectin, there were significantly increased (from 37.5 ± 9.36 to 48.8 ± 7.38 ngnml1, p < 0.05) compared to baseline. Although it remains to be confirmed with larger group of volunteers, this pilot study has demonstrated that daily consumption of banana (@ 250 g/day) is harmless both in diabetic and hypercholesterolemic volunteers and marginally beneficial to the later.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Natriumfluorid, ReagentPlus®, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Supelco
Natriumfluorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumfluorid, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Natriumfluorid, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Natriumfluorid, BioXtra, ≥99%
Sigma-Aldrich
Natriumfluoridlösung
Sigma-Aldrich
Natriumfluorid, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumfluorid 0,5 M -Lösung
Supelco
Fluorid-Ionenlösung für ISE, 0.1 M F-, for ion-selective electrodes
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.