Merck
  • Isolation and characterization of feruloylated arabinoxylan oligosaccharides from the perennial cereal grain intermediate wheat grass (Thinopyrum intermedium).
Isolation and characterization of feruloylated arabinoxylan oligosaccharides from the perennial cereal grain intermediate wheat grass (Thinopyrum intermedium).
Carbohydrate research (2015-02-24)
Rachel R Schendel, Andreas Becker, Catrin E Tyl, Mirko Bunzel
ZUSAMMENFASSUNG
In comparison to the annual grain crops dominating current agricultural production, perennial grain species require fewer chemical and energy inputs and improve soil health and erosion control. The possibility for producing sustainable grain harvests from marginal land areas is motivating research initiatives to integrate perennial grains into commercial cropping and food processing systems. In this study, the feruloylated arabinoxylans from intermediate wheat grass (Thinopyrum intermedium, IWG), a promising perennial grain candidate in agronomic screening studies, were investigated. Insoluble fiber isolated from IWG whole grain flour was subjected to either mildly acidic (50 mM TFA, 100 °C, 2 h) or enzymatic (Driselase) hydrolysis. The liberated feruloylated arabinoxylan oligosaccharides were concentrated with Amberlite XAD-2, separated with gel chromatography (Sephadex LH-20, water), and purified with reversed-phase HPLC (C18, water-MeOH gradient). Thirteen feruloylated oligosaccharides were isolated (including eight structures described for the first time) and identified by LC-ESI-MS and NMR. Linkage-type analysis via methylation analysis, as well as the monosaccharide and phenolic acid profiles of the IWG insoluble fiber were also determined. IWG feruloylated arabinoxylans have a relatively simple structure with only short feruloylated side chains, a lower backbone substitution rate than annual rye and wheat varieties, and a moderate phenolic acid content.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Essigsäureanhydrid, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Methylimidazol, ReagentPlus®, 99%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Iodmethan, contains copper as stabilizer, ReagentPlus®, 99%