Merck
  • Metabolic flux pattern of glucose utilization by Xanthomonas campestris pv. campestris: prevalent role of the Entner-Doudoroff pathway and minor fluxes through the pentose phosphate pathway and glycolysis.
Metabolic flux pattern of glucose utilization by Xanthomonas campestris pv. campestris: prevalent role of the Entner-Doudoroff pathway and minor fluxes through the pentose phosphate pathway and glycolysis.
Molecular bioSystems (2014-07-30)
Sarah Schatschneider, Claudia Huber, Heiko Neuweger, Tony Francis Watt, Alfred Pühler, Wolfgang Eisenreich, Christoph Wittmann, Karsten Niehaus, Frank-Jörg Vorhölter
ZUSAMMENFASSUNG
The well-studied plant pathogenic bacterium Xanthomonas campestris pv. campestris (Xcc) synthesizes the biotechnologically important polysaccharide xanthan gum, which is also regarded as a virulence factor in plant interactions. In Xcc, sugars like glucose are utilized as a source to generate energy and biomass for growth and pathogenicity. In this study, we used [1-(13)C]glucose as a tracer to analyze the fluxes in the central metabolism of the bacterium growing in a minimal medium. (13)C-Metabolic flux analysis based on gas chromatography-mass spectrometry (GC-MS) confirmed the prevalent catabolic role of the Entner-Doudoroff pathway. Comparative nuclear magnetic resonance (NMR)-based isotopologue profiling of a mutant deficient in glycolysis gave evidence for a moderate flux via glycolysis in the wild-type. In addition to reconfirming the Entner-Doudoroff pathway as a catabolic main route, this approach affirmed a numerically minor but important flux via the pentose phosphate pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Supelco
Dimethylformamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Supelco
N,N-Dimethylformamid, analytical standard
Supelco
Pyridin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyridin, analytical standard