Merck
  • Estrogen receptor beta as a novel target of androgen receptor action in breast cancer cell lines.
Estrogen receptor beta as a novel target of androgen receptor action in breast cancer cell lines.
Breast cancer research : BCR (2014-02-21)
Pietro Rizza, Ines Barone, Domenico Zito, Francesca Giordano, Marilena Lanzino, Francesca De Amicis, Loredana Mauro, Diego Sisci, Stefania Catalano, Karin Dahlman Wright, Jan-Ake Gustafsson, Sebastiano Andò
ZUSAMMENFASSUNG
The two isoforms of estrogen receptor (ER) alpha and beta play opposite roles in regulating proliferation and differentiation of breast cancers, with ER-alpha mediating mitogenic effects and ER-beta acting as a tumor suppressor. Emerging data have reported that androgen receptor (AR) activation inhibits ER-positive breast cancer progression mainly by antagonizing ER-alpha signaling. However, to date no studies have specifically evaluated a potential involvement of ER-beta in the inhibitory effects of androgens. ER-beta expression was examined in human breast cancer cell lines using real-time PCR, Western blotting and small interfering RNA (siRNA) assays. Mutagenesis studies, electromobility shift assay (EMSA) and chromatin immunoprecipitation (ChIP) analysis were performed to assess the effects of mibolerone/AR on ER-beta promoter activity and binding. In this study, we demonstrate that mibolerone, a synthetic androgen ligand, up-regulates ER-beta mRNA and protein levels in ER-positive breast cancer cells. Transient transfection experiments, using a vector containing the human ER-beta promoter region, show that mibolerone increases basal ER-beta promoter activity. Site-directed mutagenesis and deletion analysis reveal that an androgen response element (ARE), TGTTCT motif located at positions -383 and -377, is critical for mibolerone-induced ER-beta up-regulation in breast cancer cells. This occurs through an increased recruitment of AR to the ARE site within the ER-beta promoter region, along with an enhanced occupancy of RNA polymerase II. Finally, silencing of ER-beta gene expression by RNA interference is able to partially reverse the effects of mibolerone on cell proliferation, p21 and cyclin D1 expression. Collectively, these data provide evidence for a novel mechanism by which activated AR, through an up-regulation of ER-beta gene expression, inhibits breast cancer cell growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
1H-1,2,3-Triazol, 97%
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
Dimethylsulfoxid, analytical standard