- Steroidal glycosides from the underground parts of Dracaena thalioides and their cytotoxic activity.
Steroidal glycosides from the underground parts of Dracaena thalioides and their cytotoxic activity.
Phytochemistry (2014-08-26)
Liying Tang, Zhuju Wang, Hongwei Wu, Akihito Yokosuka, Yoshihiro Mimaki
PMID25152452
ZUSAMMENFASSUNG
Six spirostanol glycosides (1-6) and 12 known compounds (7-18) were isolated from the underground parts of Dracaena thalioides (Agavaceae). Their structures were determined by spectroscopic analysis, including 2D NMR spectroscopic data, and chemical transformations. The isolated compounds were evaluated for cytotoxic activity against HL-60 human leukemia cells. Compounds 1, 3-6, and 8-18 showed cytotoxicity against HL-60 cells, of which 10, a bisdesmosidic spirostanol derivative, showed potent cytotoxicity against HL-60 cells with an IC50 value of 0.38μM and induced apoptosis in HL-60 cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioXtra, suitable for mouse embryo cell culture
Streptomycinsulfat, European Pharmacopoeia (EP) Reference Standard