Merck
  • Capsaicin causes inactivation and degradation of the androgen receptor by inducing the restoration of miR-449a in prostate cancer.
Capsaicin causes inactivation and degradation of the androgen receptor by inducing the restoration of miR-449a in prostate cancer.
Oncology reports (2015-06-18)
Long Zheng, Jiaqi Chen, Zhenkun Ma, Wei Liu, Fei Yang, Zhao Yang, Ke Wang, Xinyang Wang, Dalin He, Lei Li
ZUSAMMENFASSUNG
Capsaicin, a novel antitumor agent extracted from chili peppers, has been proven to induce growth inhibition in various types of cancer including prostate cancer. However, the detailed mechanisms remain largely undiscovered. In the present study, we explored the regulation of the androgen receptor (AR) by capsaicin and further researched the mechanisms of their interaction in AR-positive prostate cancer cells. In the present study, cell viability was assessed by MTT assay. Cell cycle distribution was determined using flow cytometry. Expression levels of cyclin D1, miR-449a, AR and prostate-specific antigen (PSA) were assessed by quantitative real-time polymerase chain reaction or western blot analysis. To further confirm the relationship among miR-449a, AR and prostate cancer proliferation, miR-449a was overexpressed by a lentivirus in prostate cancer cells. We discovered that capsaicin prevented tumor proliferation and cell cycle progression through inactivation and degradation of AR. We also found that restoration of miR-449a induced by capsaicin treatment resulted in the inhibition of AR signaling. Finally, we demonstrated that increased expression of miR-449a sensitized prostate cancer to capsaicin treatment. Finally, our experimental results indicated that capsaicin negatively modulates the activity of AR at the mRNA and protein levels by restoring miR-449a profiling in prostate cancer. In addition, increased expression of miR-449a may facilitate the sensitivity of prostate cancer to capsaicin treatment. Thus, capsaicin may be developed as a novel anti-AR drug for the therapy of prostate cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®