Merck
  • JTP-103237, a monoacylglycerol acyltransferase inhibitor, prevents fatty liver and suppresses both triglyceride synthesis and de novo lipogenesis.
JTP-103237, a monoacylglycerol acyltransferase inhibitor, prevents fatty liver and suppresses both triglyceride synthesis and de novo lipogenesis.
Journal of pharmacological sciences (2015-07-29)
Chihiro Okuma, Takeshi Ohta, Hironobu Tadaki, Tatsuya Ishigure, Shohei Sakata, Hideyuki Taniuchi, Ryuhei Sano, Hiromi Hamada, Shinichi Kume, Jun Nishiu, Makoto Kakutani
ZUSAMMENFASSUNG
Monoacyglycerol acyltransferases (MGATs) are known to play important roles in intestinal TG absorption. In contrast, the role of MGATs in the liver is still unclear. We investigated the effects of JTP-103237, a novel MGAT inhibitor, on hepatic MGAT activity and hepatic lipid metabolism. JTP-103237 reduced hepatic triglyceride content and hepatic MGAT activity in a high sucrose very low fat (HSVLF) diet induced fatty liver model. Interestingly, JTP-103237 suppressed not only triglyceride (TG) and diacylglycerol (DG) synthesis, but also fatty acid (FA) synthesis (de novo lipogenesis) in this model. JTP-103237 also suppressed lipogenesis-related gene expression, such as sterol regulatory element-binding protein 1-c. Moreover, JTP-103237 decreased plasma glucose levels and total cholesterol and reduced the accumulation of epididymal fats in HSVLF diet fed mice. In the present study, JTP-103237 prevented carbohydrate-induced fatty liver and suppressed both TG synthesis and de novo lipogenesis, suggesting MGAT inhibitor may prevent carbohydrate-induced metabolic disorders, including NAFLD, obesity and diabetes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Palmitinsäure, ≥99%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, suitable for cell culture, BioReagent
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Palmitinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Palmitinsäure, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitinsäure, natural, 98%, FG
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)