Merck
  • A method for studies on interactions between a gold-based drug and plasma proteins based on capillary electrophoresis with inductively coupled plasma mass spectrometry detection.
A method for studies on interactions between a gold-based drug and plasma proteins based on capillary electrophoresis with inductively coupled plasma mass spectrometry detection.
Analytical and bioanalytical chemistry (2015-09-04)
Tam T T N Nguyen, Jesper Østergaard, Bente Gammelgaard
ZUSAMMENFASSUNG
An analytical method based on capillary electrophoresis (CE) and inductively coupled plasma mass spectrometry (ICP-MS) detection was developed for studies on the interaction of gold-containing drugs and plasma proteins using auranofin as example. A detection limit of 18 ng/mL of auranofin corresponding to 5.2 ng/mL Au and a precision of 1.5 % were obtained. Kinetic studies of the interaction between auranofin and protein were performed by incubation in aqueous solutions as well as 20 % human plasma at 37 °C. The reaction of auranofin with human serum albumin (HSA) and plasma proceeded fast; 50 % of un-bound auranofin disappeared within 2 and 3 min, respectively. By blocking the free cysteine (Cys-34) by iodoacetamide on HSA, it was shown that Cys-34 was the main reaction site for auranofin. By selective labeling of HSA present in 20 % human plasma with iophenoxate, it was demonstrated that HSA was the major auranofin-interacting protein in plasma. The CE-ICP-MS method is proposed as a novel approach for kinetic studies of the interactions between gold-based drugs and plasma proteins. Graphical Abstract Development of a CE-ICP-MS based method allows for studies on interaction of the gold containing drug auranofin with plasma proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Auranofin, ≥98% (HPLC)
SAFC
Jodacetamid
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.