Merck
Increased production of intestinal immunoglobulins in Syntenin-1-deficient mice.
Immunobiology (2014-12-30)
Kentaro Tamura, Masashi Ikutani, Taketoshi Yoshida, Ayumi Tanaka-Hayashi, Tsutomu Yanagibashi, Ran Inoue, Yoshinori Nagai, Yuichi Adachi, Toshio Miyawaki, Kiyoshi Takatsu, Hisashi Mori
ZUSAMMENFASSUNG
Syntenin-1 is an intracellular PDZ protein that binds multiple proteins and regulates protein trafficking, cancer metastasis, exosome production, synaptic formation, and IL-5 signaling. However, the functions of Syntenin-1 have not yet been clearly characterized in detail, especially in vivo. In this study, we generated a Syntenin-1 knock out (KO) mouse strain and analyzed the role(s) of Syntenin-1 in IL-5 signaling, because the direct interaction of Syntenin-1 with the cytoplasmic domain of the IL-5 receptor α subunit and the regulation of IL-5 signaling by Syntenin-1 have been reported. Unexpectedly, the number of IL-5-responding cells was normal and the levels of fecal immunoglobulins were rather higher in the Syntenin-1 KO mice. We also found that IgA and IgM production of splenic B cells stimulated in vitro was increased in Syntenin-1 KO mice. In addition, we showed that a distribution of intestinal microbial flora was influenced in Syntenin-1 KO mice. Our data indicate that Syntenin-1 negatively regulates the intestinal immunoglobulin production and has a function to maintain the intestinal homeostasis in vivo. The analysis of Syntenin-1 KO mice may provide novel information on not only mucosal immunity but also other functions of Syntenin-1 such as cancer metastasis and neural development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Ammoniumhydrogendifluorid, 99.999% trace metals basis
Supelco
N,N′-Bis(acryloyl)cystamin, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)