Merck
  • Human papillomavirus 16-specific cell-mediated immunity in children born to mothers with incident cervical intraepithelial neoplasia (CIN) and to those constantly HPV negative.
Human papillomavirus 16-specific cell-mediated immunity in children born to mothers with incident cervical intraepithelial neoplasia (CIN) and to those constantly HPV negative.
Journal of translational medicine (2015-11-27)
Hanna-Mari Koskimaa, Anna Paaso, Marij J P Welters, Seija Grénman, Kari Syrjänen, Sjoerd H van der Burg, Stina Syrjänen
ZUSAMMENFASSUNG
HPV infections are detected in sexually naive children. This has raised the question about the role of early HPV infections in either protecting or predisposing to further HPV infections. HPV16-specific cell-mediated immunity (CMI) was studied in 10 case-children born to mothers with an incident cervical intraepithelial neoplasia (CIN) diagnosed during their 14-year follow-up (FU), and in 21 children born to mothers, who remained constantly HPV-negative (controls). The mean age of children was 12.3 years. Peripheral blood mononuclear cells were isolated from blood and stimulated with peptide pools covering HPV16 E2, E6 and E7. Proliferation of lymphocytes, their secretion of cytokines, and the frequency of regulatory T-cells were determined. The results were correlated with the HPV status and analyzed in a nested case-control setting. All children, except two controls, displayed CMI against HPV16 E2, E6 and/or E7 peptides associated with type 1 and 2 cytokine secretion. Only two statistically significant differences were found in the nested case-control setting; (1) case-children had a higher TNF-α response to HPV16 E2 (p = 0.004) than controls and (2) controls had no response to HPV16 E7.2 peptide pool while 3/10 case-children had (p = 0.013). Totally, 50 and 57 % of the cases and controls, respectively, had HPV positive oral samples at some FU-visit. In addition, the children without any HPV antibodies before the age of 6 months showed proliferative responses of PBMC after HPV16 exposure more frequently than other children (p = 0.045). HPV16-specific CMI is common in young, sexually inexperienced children. This suggests that oral HPV infections occur frequently in children. Our results might also explain the previous findings that half of healthy adults demonstrate HPV-specific CMI irrespective of their partner/sexual status.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®