Merck
The role of Cas-L/NEDD9 as a regulator of collagen-induced arthritis in a murine model.
Biochemical and biophysical research communications (2015-04-08)
Tomoki Katayose, Satoshi Iwata, Naoki Oyaizu, Osamu Hosono, Taketo Yamada, Nam H Dang, Ryo Hatano, Hirotoshi Tanaka, Kei Ohnuma, Chikao Morimoto
ZUSAMMENFASSUNG
Cas-L/NEDD9 is a cytoplasmic docking protein downstream of β1 integrin-mediated signaling pathway and is essential for cellular migration and β1 integrin-mediated costimulation of T cells. We previously found that increased number of Cas-L positive leukocytes migrated into the inflamed joints of HTLV-I tax transgenic mice which spontaneously develop polyarthritis, suggesting a role of Cas-L in rheumatoid arthritis (RA) pathophysiology. Our current study expanded these findings on the role of Cas-L/NEDD9 in the development of RA by analyzing the pathophysiological changes in a Nedd9(-/-) mouse collagen-induced arthritis (CIA) model. Nedd9(-/-) mice exhibited a decrease in arthritis severity as compared to Nedd9(+/+) mice. In addition, as being conducted bone marrow transplantation experiments with a CIA model, Nedd9(-/-)→Nedd9(+/+) transplant showed a decrease in the incidence and severity score of arthritis, compared to those of Nedd9(+/+)→Nedd9(-/-) transplant. For analysis of serum levels of various cytokines, IL-1β, IL-6, IL-17, TNF-α, IFN-γ and anti-collagen antibody were decreased, while IL-4 and IL-10 levels were increased, in Nedd9(-/-) mice as compared to those in Nedd9(+/+) mice. Furthermore, collagen-mediated cellular responses of lymphocytes isolated from spleen or affected lymph nodes of Nedd9(-/-) mice were reduced. Our results strongly suggest that Cas-L/NEDD9 plays a pivotal role in the pathophysiology of CIA, and that Cas-L/NEDD9 may be a potential molecular target for the treatment of RA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C