Merck
  • Simultaneous analysis of multiple neurotransmitters by hydrophilic interaction liquid chromatography coupled to tandem mass spectrometry.
Simultaneous analysis of multiple neurotransmitters by hydrophilic interaction liquid chromatography coupled to tandem mass spectrometry.
Journal of chromatography. A (2015-04-15)
Sara Tufi, Marja Lamoree, Jacob de Boer, Pim Leonards
ZUSAMMENFASSUNG
Neurotransmitters are endogenous metabolites that allow the signal transmission across neuronal synapses. Their biological role is crucial for many physiological functions and their levels can be changed by several diseases. Because of their high polarity, hydrophilic interaction liquid chromatography (HILIC) is a promising tool for neurotransmitter analysis. Due to the large number of HILIC stationary phases available, an evaluation of the column performances and retention behaviors has been performed on five different commercial HILIC packing materials (silica, amino, amide and two zwitterionic stationary phases). Several parameters like the linear correlation between retention and the distribution coefficient (logD), the separation factor k and the column resolution Rs have been investigated and the column performances have been visualized with a heat map and hierarchical clustering analysis. An optimized and validated HILIC-MS/MS method based on the ZIC-cHILIC column is proposed for the simultaneous detection and quantification of twenty compounds consisting of neurotransmitters, precursors and metabolites: 3-methoxytyramine (3-MT), 5-hydroxyindoleacetic acid (5-HIAA), 5-hydroxy-L-tripthophan, acetylcholine, choline, L-3,4-dihydroxyphenylalanine (L-DOPA), dopamine, epinephrine, γ-aminobutyric acid (GABA), glutamate, glutamine, histamine, histidine, L-tryptophan, L-tyrosine, norepinephrine, normetanephrine, phenylalanine, serotonin and tyramine. The method was applied to neuronal metabolite profiling of the central nervous system of the freshwater snail Lymnaea stagnalis. This method is suitable to explore neuronal metabolism and its alteration in different biological matrices.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
L-Tyrosin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Histamin, ≥97.0%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Tyramin, ≥98.0%
Sigma-Aldrich
L-Tyrosin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Natriumformiat, BioUltra, ≥99.0% (NT)
SAFC
L-Tyrosin
Sigma-Aldrich
DL-Phenylalanin, ReagentPlus®, 99%
Sigma-Aldrich
Natriumformiat, 99.998% trace metals basis
Sigma-Aldrich
3-Methoxytyramin -hydrochlorid, 99% (AT)
Sigma-Aldrich
L-Tyrosin, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Tyramin, 98%, FG
Sigma-Aldrich
3-Methoxytyramin -hydrochlorid, ≥95.5%, crystalline