Merck
  • Comparisons of urinary phthalate metabolites and daily phthalate intakes among Japanese families.
Comparisons of urinary phthalate metabolites and daily phthalate intakes among Japanese families.
International journal of hygiene and environmental health (2015-04-19)
Yu Ait Bamai, Atsuko Araki, Toshio Kawai, Tazuru Tsuboi, Eiji Yoshioka, Ayako Kanazawa, Shi Cong, Reiko Kishi
ZUSAMMENFASSUNG
We measured urinary phthalate metabolites, including di-n-butyl phthalate (DnBP), di-isobutyl phthalate, benzyl butyl phthalate (BBzP), and di(2-ethylhexyl) phthalate (DEHP), from 178 school-aged children and their 284 family members using gas chromatography-mass spectrometry, and we calculated daily phthalate intakes. The highest median levels of phthalate metabolites were for mono-isobutyl phthalate in all participants except schoolchildren, where the highest levels were for mono-(2-ethyl-5-oxohexyl) phthalate (MEOHP). Comparing the schoolchildren with their parents, the schoolchildren had significantly higher urinary metabolites for MEOHP, mono-(2-ethyl-5-carboxypentyl) phthalate, and ΣDEHP. Regarding daily intakes, the schoolchildren had significantly higher daily intakes of DnBP, BBzP, and ΣDEHP. All phthalate metabolite and sums of metabolite levels in the schoolchildren were positively correlated with their mothers' levels, except for MEHP, whereas fathers were less correlated with their children. The DEHP intake in this study was higher than that of most other studies. Moreover, 10% of the children and 3% of the adults exceeded the Reference Dose (RfD) value (20μg/kg/day) of the U.S. Environmental Protection Agency, which indicates that it is important to focus on children's DEHP exposure because the children exceeded the RfD more than adults among the same families who shared similar exposure sources. Our results will contribute to considerations of the regulations for some phthalates and the actual phthalate exposure levels in the Japanese population.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Natriumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Natriumsulfat, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Natriumsulfat, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C