Merck
  • Structure-Activity and Structure-Property Relationship and Exploratory in Vivo Evaluation of the Nanomolar Keap1-Nrf2 Protein-Protein Interaction Inhibitor.
Structure-Activity and Structure-Property Relationship and Exploratory in Vivo Evaluation of the Nanomolar Keap1-Nrf2 Protein-Protein Interaction Inhibitor.
Journal of medicinal chemistry (2015-08-11)
Zheng-Yu Jiang, Li Li Xu, Meng-Chen Lu, Zhi-Yun Chen, Zhen-Wei Yuan, Xiao-Li Xu, Xiao-Ke Guo, Xiao-Jin Zhang, Hao-Peng Sun, Qi-Dong You
ZUSAMMENFASSUNG
Directly disrupting the Keap1-Nrf2 protein-protein interaction (PPI) is an effective way to activate Nrf2. Using the potent Keap1-Nrf2 PPI inhibitor that was reported by our group, we conducted a preliminary investigation of the structure-activity and structure-property relationships of the ring systems to improve the drug-like properties. Compound 18e, which bore p-acetamido substituents on the side chain phenyl rings, was the best choice for balancing PPI inhibition activity, physicochemical properties, and cellular Nrf2 activity. Cell-based experiments with 18e showed that the Keap1-Nrf2 PPI inhibitor can activate Nrf2 and induce the expression of Nrf2 downstream proteins in an Nrf2-dependent manner. An exploratory in vivo experiment was carried out to further evaluate the anti-inflammatory effects of 18e in a LPS-challenged mouse model. The primary results indicated that 18e could reduce the level of circulating pro-inflammatory cytokines induced by LPS and relieve the inflammatory response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by trichloroacetic acid extraction
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
SAFC
McCoy 5A-Medium, Modified, with L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Tetramethylsilan, ≥99.0% (GC)
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Tetramethylsilan, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nfe2l2