Merck
Physiological and Molecular Responses to Excess Boron in Citrus macrophylla W.
PloS one (2015-08-01)
Mary-Rus Martínez-Cuenca, Belén Martínez-Alcántara, Ana Quiñones, Marta Ruiz, Domingo J Iglesias, Eduardo Primo-Millo, M Ángeles Forner-Giner
ZUSAMMENFASSUNG
This work provides insight into several mechanisms involved in boron (B) regulation pathway in response to high B conditions in Citrus. The study was carried out in Citrus macrophylla W. (Cm) seedlings cultured "in vitro" in media with 50 or 400 μM H3BO3 (control, Ct, and B-excess, +B, plants, respectively). Growth parameters, B concentration, leaf chlorophyll (Chl) concentration, the expression of the main putative genes involved in B transport and distribution, and leaf and root proline and malonaldehyde (MDA) concentrations, were assessed. Excess B led to high B concentration in +B plants (3.8- and 1.4-fold in leaves and roots, respectively) when compared with Ct ones. However, a minor effect was recorded in the plant (incipient visual symptoms, less than 27% reduction in root growth and 26% decrease in Chl b concentration). B toxicity down-regulated by half the expression level of putative B transporter genes NIP5 and PIP1. CmBOR1 gene was not repressed in +B plants and B accumulated in the shoots. High B level increased the transcripts of putative gene TIP5, involved in B transport across the tonoplast, by 3.3- and 2.4-fold in leaves and roots, respectively. The activity of V-PPiase proton pump, related with the electrochemical gradient in the vacuole, was also enhanced in +B organs. B toxicity up-regulated putative BOR4 gene (2.1- and 2.7-fold in roots and leaves, respectively), which codifies for an active efflux B transporter. Accordingly, B was located in +B plants preferently in an insoluble form on cell walls. Finally, excess B caused a significant rise in proline concentration (51% and 34% in roots and leaves, respectively), while the MDA level did not exceed 20%. In conclusion, Cm tolerance to a high B level is likely based on the synergism of several specific mechanisms against B toxicity, including: 1/ down-regulation of NIP5 and PIP1 boron transporters; 2/ activation of B efflux from cells due to the up-regulation of putative BOR4 gene; 3/ compartmentation of B in the vacuole through TIP5 transporter activation and the acidification of the organelle; 4/ insolubilisation of B and deposition in cell walls preventing from cytoplasm damage; and, 5/ induction of an efficient antioxidant system through proline accumulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Natriumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Natriumsulfat, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Natriumsulfat, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O