Merck
  • Organ-protective effects on the liver and kidney by minocycline in small piglets undergoing cardiopulonary bypass.
Organ-protective effects on the liver and kidney by minocycline in small piglets undergoing cardiopulonary bypass.
Naunyn-Schmiedeberg's archives of pharmacology (2015-03-17)
Stefan Dhein, Maria Grassl, Maria Gerdom, Marcel Vollroth, Farhad Bakhtiary, Sandy von Salisch, Klaus Krämer, Axel Sobiraj, Martin Kostelka, Friedrich-Wilhelm Mohr, Aida Salameh
ZUSAMMENFASSUNG
Cardiopulmonary bypass (CPB) often is required for the operative correction of congenital heart defects in small infants. Unfortunately, CPB is associated with injury of inner organs such as the brain, kidney, lung, and liver. Renal failure and increase in liver enzymes are typical side effects observed after CPB. Here, we investigate whether organ protection of the kidney and liver can be achieved with the application of minocycline, which is known-besides its anti-infective effects-to act as a poly-ADP-ribose-polymerase inhibitor. Twenty-nine 4-week-old Angler Sattelschwein-piglets (8-15 kg) were divided into four groups: control group (n = 8), CPB group (n = 9), minocycline-control group (n = 6), and the minocycline-CPB group (n = 6). CPB groups were thoracotomized and underwent CPB for 120 min (cross-clamp, 90 min; reperfusion, 30 min) followed by a 90-min recovery time. The control groups also were thoracotomized but not connected to CPB. The minocycline group received 4 mg/kg minocycline before and 2 mg/kg after CPB. In the kidneys, CPB histologically resulted in widening of Bowman's capsule, and-mainly in tubules-formation of poly-ADP-ribose, nitrosylation of tyrosine-residues, nuclear translocation of hypoxia-induced factor HIF-1α, and of apoptosis-inducing factor (AIF). In addition, we found significantly less ATP in the kidney and significantly increased plasma urea and creatinine. Similar but gradually attenuated changes were found in the liver together with significantly elevated de-Ritis coefficient. These changes in the kidney and liver were significantly diminished by minocycline (except AIF in the liver which was similar in all groups). In conclusion, CPB causes damage in the kidney and-to a lower degree-in the liver, which can be attenuated by minocycline.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
3-Amino-9-ethylcarbazol, tablet
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
3-Amino-9-ethylcarbazol, ≥95% (HPLC), powder
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
9-Ethylcarbazol, 97%
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis