Merck
  • Development of Transcriptomic Markers for Population Analysis Using Restriction Site Associated RNA Sequencing (RARseq).
Development of Transcriptomic Markers for Population Analysis Using Restriction Site Associated RNA Sequencing (RARseq).
PloS one (2015-08-05)
Magdy S Alabady, Willie L Rogers, Russell L Malmberg
ZUSAMMENFASSUNG
We describe restriction site associated RNA sequencing (RARseq), an RNAseq-based genotype by sequencing (GBS) method. It includes the construction of RNAseq libraries from double stranded cDNA digested with selected restriction enzymes. To test this, we constructed six single- and six-dual-digested RARseq libraries from six F2 pitcher plant individuals and sequenced them on a half of a Miseq run. On average, the de novo approach of population genome analysis detected 544 and 570 RNA SNPs, whereas the reference transcriptome-based approach revealed an average of 1907 and 1876 RNA SNPs per individual, from single- and dual-digested RARseq data, respectively. The average numbers of RNA SNPs and alleles per loci are 1.89 and 2.17, respectively. Our results suggest that the RARseq protocol allows good depth of coverage per loci for detecting RNA SNPs and polymorphic loci for population genomics and mapping analyses. In non-model systems where complete genomes sequences are not always available, RARseq data can be analyzed in reference to the transcriptome. In addition to enriching for functional markers, this method may prove particularly useful in organisms where the genomes are not favorable for DNA GBS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O