Merck
  • Effect of Probiotic Supplement on Cytokine Levels in HIV-Infected Individuals: A Preliminary Study.
Effect of Probiotic Supplement on Cytokine Levels in HIV-Infected Individuals: A Preliminary Study.
Nutrients (2015-10-02)
Katia Falasca, Jacopo Vecchiet, Claudio Ucciferri, Marta Di Nicola, Chiara D'Angelo, Marcella Reale
ZUSAMMENFASSUNG
Inflammation persists in patients infected with HIV. Reduction of inflammatory cytokines and microbial translocation might be one way that this could be managed. The anti-inflammatory properties of certain probiotic strains prompted us to investigate whether a probiotic could reduce the inflammatory index of HIV-infected patients. The study involved 30 HIV+ males on antiretroviral therapy, who were given one bottle of fermented milk Yakult Light® containing Lactobacillus casei Shirota (LcS) twice a day for four weeks. The probiotic LcS was associated with an increase of T lymphocytes and a significant increase of CD56+ cells (p = 0.04). There was also a significant decrease of mRNA levels of TGFβ, IL-10 and IL-12 (p < 0.001) and IL-1β expression (p < 0.001) and an increase of serum IL-23 (p = 0.03). In addition, decreased inflammation and cardiovascular risk were observed, as shown by a reduction of cystatin C (p < 0.001). These data provide preliminary evidence that probiotic supplementation may modulate certain immunological parameters and some of the cytokines that were analyzed. Thus, we propose that LcS may be an inexpensive and practical strategy to support the immune function of HIV+ patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%