Merck
  • Exploring the isoform selectivity of TGX-221 related pyrido[1,2-a]pyrimidinone-based Class IA PI 3-kinase inhibitors: synthesis, biological evaluation and molecular modelling.
Exploring the isoform selectivity of TGX-221 related pyrido[1,2-a]pyrimidinone-based Class IA PI 3-kinase inhibitors: synthesis, biological evaluation and molecular modelling.
Bioorganic & medicinal chemistry (2015-04-22)
Andrew J Marshall, Claire L Lill, Mindy Chao, Sharada V Kolekar, Woo-Jeong Lee, Elaine S Marshall, Bruce C Baguley, Peter R Shepherd, William A Denny, Jack U Flanagan, Gordon W Rewcastle
ZUSAMMENFASSUNG
A novel series of TGX-221 analogues was prepared and tested for their potency against the p110α, p110β, and p110δ isoforms of the PI3K enzyme, and in two cellular assays. The biological results were interpreted in terms of a p110β comparative model, in order to account for their selectivity towards this isoform. A CH2NH type linker is proposed to allow binding into the specificity pocket proposed to accommodate the high p110β-selectivity of TGX-221, although there was limited steric tolerance for substituents on the pendant ring with the 2-position most favourable for substitution.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Nitrilotriessigsäure, Sigma Grade, ≥99%
Sigma-Aldrich
Ammoniumpersulfat, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Nitrilotriessigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammoniumpersulfat, BioXtra, ≥98.0%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Nitrilotriessigsäure, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®