Merck
  • Rho-kinase inhibitor Y-27632 increases cellular proliferation and migration in human foreskin fibroblast cells.
Rho-kinase inhibitor Y-27632 increases cellular proliferation and migration in human foreskin fibroblast cells.
Proteomics (2015-05-08)
Juha Piltti, Markku Varjosalo, Chengjuan Qu, Jukka Häyrinen, Mikko J Lammi
ZUSAMMENFASSUNG
The idea of direct differentiation of somatic cells into other differentiated cell types has attracted a great interest recently. Rho-kinase inhibitor Y-27632 (ROCKi) is a potential drug molecule, which has been reported to support the gene expressions typical for the chondrocytes, thus restricting their phenotypic conversion to fibroblastic cells upon the cellular expansion. In this study, we have investigated the short-term biological responses of ROCKi to human primary foreskin fibroblasts. The fibroblast cells were exposed to 1 and 10 μM ROCKi treatments. A proteomics analysis revealed expression changes of 56 proteins, and a further protein pathway analysis suggested their association with the cell morphology, the organization, and the increased cellular movement and the proliferation. These functional responses were confirmed by a Cell-IQ time-lapse imaging analysis. Rho-kinase inhibitor treatment increased the cellular proliferation up to twofold during the first 12 h, and a wound model based migration assay showed 50% faster filling of the mechanically generated wound area. Additionally, significantly less vinculin-associated focal adhesions were present in the ROCKi-treated cells. Despite the marked changes in the cell behavior, ROCKi was not able to induce the expression of the chondrocyte-specific genes, such as procollagen α1 (II) and aggrecan.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
SAFC
L-Glutamin
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C