Merck
Assessment of arsenic bioaccessibility in raw and cooked edible mushrooms by a PBET method.
Food chemistry (2015-10-17)
Toni Llorente-Mirandes, Mariona Llorens-Muñoz, Virginia Funes-Collado, Àngels Sahuquillo, José Fermín López-Sánchez
ZUSAMMENFASSUNG
The present study reports arsenic analysis in Lentinula edodes, Agaricus bisporus and Pleurotus ostreatus before and after being cooked. Furthermore, arsenic in raw and cooked mushroom was determined in the gastric and gastrointestinal bioaccessible fractions obtained after simulating human digestion by means of an in vitro physiologically based extraction test (PBET). Several certified reference materials (SRM 1568a, SRM 1570a, CRM 7503-a, BC211 and IPE-120) were analysed to evaluate the proposed methods. Total arsenic content was 1393, 181 and 335μgAskg(-1) for L. edodes, A. bisporus and P. ostreatus, respectively, and decreased by between 53% and 71% in boiled mushroom and less than 11% in griddled mushroom. High bioaccessibility was observed in raw, boiled and griddled mushroom, ranging from 74% to 89% and from 80% to 100% for gastric and gastrointestinal extracts, respectively, suggesting the need to consider the potential health risk of consumption of the mushrooms analysed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Maleinsäure, ReagentPlus®, ≥99% (HPLC)