Merck
  • Supercritical CO₂ extraction of oil, fatty acids and flavonolignans from milk thistle seeds: Evaluation of their antioxidant and cytotoxic activities in Caco-2 cells.
Supercritical CO₂ extraction of oil, fatty acids and flavonolignans from milk thistle seeds: Evaluation of their antioxidant and cytotoxic activities in Caco-2 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2015-07-15)
Naila Ben Rahal, Francisco J Barba, Danielle Barth, Isabelle Chevalot
ZUSAMMENFASSUNG
The optimal conditions of supercritical carbon dioxide (SC-CO2) (160-220 bars, 40-80 °C) technology combined with co-solvent (ethanol), to recover oil, flavonolignans (silychristin, silydianin and silybinin) and fatty acids from milk thistle seeds, to be used as food additives and/or nutraceuticals, were studied. Moreover, the antioxidant and cytotoxic activities of the SC-CO2 oil seeds extracts were evaluated in Caco-2 carcinoma cells. Pressure and temperature had a significant effect on oil and flavonolignans recovery, although there was not observed a clear trend. SC-CO2 with co-solvent extraction at 220 bars, 40 °C was the optimum treatment to recover oil (30.8%) and flavonolignans from milk thistle seeds. Moreover, linoleic (47.64-66.70%), and oleic (19.68-24.83%) acids were the predominant fatty acids in the oil extracts recovered from milk thistle under SC-CO2. In addition, SC-CO2 extract showed a high antioxidant activity determined by DPPH and ABTS tests. Cytotoxic activities of silychristin, silydianin and silybinin and the obtained SC-CO2 extract (220 bars, 40 °C) were evaluated against Caco-2 cells. The SC-CO2 extract inhibited the proliferation of Caco-2 cells in a dose-responsive manner and induced the highest percentage of mortality of Caco-2 cells (from 43 to 71% for concentrations from 10 up to 100 μg/ml of SC-CO2 oil seeds).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin