Merck
Push-out bond strength of CPP-ACP-modified calcium silicate-based cements.
Dental materials journal (2015-08-04)
Alaa E Dawood, David J Manton, Peter Parashos, Rebecca H k Wong, Joseph E A Palamara, Eric C Reynolds
ZUSAMMENFASSUNG
This study evaluated the push-out bond strength of 0%, 0.5%, 1.0%, 2.0% and 3.0% (w/w) casein phosphopeptide-amorphous calcium phosphate (CPP-ACP)-modified calcium silicate-based cements (CSCs). The push-out bond strength of a trial MTA was also compared with two CSCs (Biodentine(™) and Angelus(®) MTA). Three hundred 1 mm thick horizontal root sections were prepared from 60 singlerooted human teeth. The canal space of each section was enlarged and filled with the cements. The sections were stored in a phosphate buffer solution. After incubation for 2 months, the push-out bond strength was measured and the data were analyzed using one way analysis of variance followed by Tukey's test. The addition of CPP-ACP to the test cements increased the push-out bond strength (p<0.05). The push-out bond strength of Biodentine(™) was higher than the other cements (p<0.05). There was no statistically significant difference between Angelus(®) MTA and the trial MTA with most of CPP-ACP concentrations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
5′-Deoxy-5′-(methylthio)adenosin