Merck
  • Enteral siRNA delivery technique for therapeutic gene silencing in the liver via the lymphatic route.
Enteral siRNA delivery technique for therapeutic gene silencing in the liver via the lymphatic route.
Scientific reports (2015-11-26)
Masahiro Murakami, Kazutaka Nishina, Chie Watanabe, Kie Yoshida-Tanaka, Wenying Piao, Hiroya Kuwahara, Yuji Horikiri, Kanjiro Miyata, Nobuhiro Nishiyama, Kazunori Kataoka, Masayuki Yoshida, Hidehiro Mizusawa, Takanori Yokota
ZUSAMMENFASSUNG
An efficient targeting delivery technology is needed for functional oligonucleotides to exert their potential effect on the target gene without an adverse effect in vivo. Development of enteral delivery systems for nucleic acids is a major challenge because of their large molecular size and instability. Here, we describe a new enteral delivery technique that enables small interfering RNA (siRNA) selectively delivered to the liver to silence its target Apolipoprotein B gene expression. A nuclease-resistant synthetic siRNA was conjugated with α-tochopherol and administered as lipid nanoparticle to the large intestine of the mice in a postprandial state. The selective transport into the liver, effective gene silence, and consequently significant reduction in serum low density lipoprotein-cholesterol level, were demonstrated. The chylomicron-mediated pathway via the lymphatic route was suggested as major mechanism. This unique approach may provide a basis for developing oral and rectal delivery systems for nucleic acids targeting liver.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
α-Linoleic acid, ≥98%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)