Merck
  • Characterization of Thiamin Phosphate Kinase in the Hyperthermophilic Archaeon Pyrobaculum calidifontis.
Characterization of Thiamin Phosphate Kinase in the Hyperthermophilic Archaeon Pyrobaculum calidifontis.
Journal of nutritional science and vitaminology (2015-12-08)
Maria Hayashi, Kazuto Nosaka
ZUSAMMENFASSUNG
Thiamin pyrophosphate is an essential cofactor in all living systems. In its biosynthesis, the thiamin structure is initially formed as thiamin phosphate from a thiazole and a pyrimidine moiety, and then thiamin pyrophosphate is synthesized from thiamin phosphate. Many eubacterial cells directly synthesize thiamin pyrophosphate by the phosphorylation of thiamin phosphate by thiamin phosphate kinase (ThiL), whereas this final step occurs in two stages in eukaryotic cells and some eubacterial cells: hydrolysis of thiamin phosphate to free thiamin and its pyrophosphorylation by thiamin pyrophosphokinase. In addition, some eubacteria have thiamin kinase, a salvage enzyme that converts the incorporated thiamin from the environment to thiamin phosphate. This final step in thiamin biosynthesis has never been experimentally investigated in archaea, although the putative thiL genes are found in their genome database. In this study, we observed thiamin phosphate kinase activity in the soluble fraction of the hyperthermophilic archaeon Pyrobaculum calidifontis. On the other hand, neither thiamin pyrophosphokinase nor thiamin kinase activity was detected, suggesting that in this archaeon the phosphorylation of thiamin phosphate is only way to synthesize thiamin pyrophosphate and it cannot use exogenous thiamin for the salvage synthesis of thiamin pyrophosphate. We also investigated the kinetic properties of thiamin phosphate kinase activity using the recombinant ThiL protein from P. calidifontis. Furthermore, the results obtained by site-directed mutagenesis suggest that the Ser196 of ThiL protein plays a pivotal role in the catalytic process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Proteasehemmer-Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Proteasehemmer-Cocktailpulver, for use with bacterial cell extracts, lyophilized powder
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O