Merck
  • Human bone marrow stem cells cultured under hypoxic conditions present altered characteristics and enhanced in vivo tissue regeneration.
Human bone marrow stem cells cultured under hypoxic conditions present altered characteristics and enhanced in vivo tissue regeneration.
Bone (2015-05-09)
Jung-Seok Lee, Jung-Chul Park, Tae-Wan Kim, Byung-Joo Jung, Youngseok Lee, Eun-Kyung Shim, Soyon Park, Eun-Young Choi, Kyoo-Sung Cho, Chang-Sung Kim
ZUSAMMENFASSUNG
Human bone marrow mesenchymal stem cells (hBMSCs) were isolated from bone marrow of the vertebral body. The hBMSCs were cultured under either hypoxic (1% O2) or normoxic (21% O2; control) conditions and the characteristics as mesenchymal stem cells were compared. Results revealed that hypoxia reduced proliferative potential and colony-forming efficiency of hBMSCs, and significantly enhanced osteogenic and chondrogenic differentiation. The hBMSCs enhanced the regenerative potential of bone in vivo. In vitro synthesis of soluble and insoluble collagen was significantly increased in the hypoxic condition. In vivo collagen tissue regeneration was also enhanced under the hypoxic condition, with concomitant increased expressions of various subtypes of collagen and lysyl-oxidase family mRNA. MicroRNA assays revealed that miR-155-5p, which negatively regulates HIF-1α, was significantly highly expressed. These observations demonstrate that hBMSCs obtained from human vertebrae exhibit altered characteristics under hypoxic conditions, and each factor contributing to hBMSC-mediated tissue healing should be evaluated with the goal of allowing their clinical application.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Hydroxyapatit, powder, synthetic
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Calciumphosphat tribasisch, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatit, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxylapatit, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxylapatit, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxylapatit, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Calciumphosphat tribasisch, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C