Merck
  • Lactoferrin and ovotransferrin contribute toward antioxidative effects of Edible Bird's Nest against hydrogen peroxide-induced oxidative stress in human SH-SY5Y cells.
Lactoferrin and ovotransferrin contribute toward antioxidative effects of Edible Bird's Nest against hydrogen peroxide-induced oxidative stress in human SH-SY5Y cells.
Bioscience, biotechnology, and biochemistry (2015-06-10)
Zhiping Hou, Mustapha Umar Imam, Maznah Ismail, Nur Hanisah Azmi, Norsharina Ismail, Aini Ideris, Rozi Mahmud
ZUSAMMENFASSUNG
There are reports of improved redox outcomes due to consumption of Edible Bird's Nest (EBN). Many of the functional effects of EBN can be linked to its high amounts of antioxidants. Interestingly, dietary components with high antioxidants have shown promise in the prevention of aging and its related diseases like Alzheimer's disease. In this study, the antioxidative potentials of EBN and its constituents, lactoferrin (LF) and ovotransferrin (OVF), were determined and protective effects against hydrogen peroxide (H2O2)- induced toxicity on SH-SY5Y cells using 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay and acridine orange and propidium iodide (AO/PI) staining with microscopy were examined. Results showed that EBN and its constituents attenuated H2O2-induced cytotoxicity, and decreased radical oxygen species (ROS) through increased scavenging activity. Furthermore, LF, OVF, and EBN produced transcriptional changes in antioxidant related genes that tended towards neuroprotection as compared to H2O2-treated group. Overall, the results suggest that LF and OVF may produce synergistic or all-or-none antioxidative effects in EBN.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
Conalbumin aus Hühnereiweiss, suitable for cell culture, BioReagent
Sigma-Aldrich
Lactoferrin aus Frauenmilch, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®