Merck
  • Analysis of expression and inhibitory activity of a TrcC-6 phytocystatin present in developing and germinating seeds of triticale (×Triticosecale Wittm.).
Analysis of expression and inhibitory activity of a TrcC-6 phytocystatin present in developing and germinating seeds of triticale (×Triticosecale Wittm.).
Plant physiology and biochemistry : PPB (2015-08-25)
Joanna Simińska, Sławomir Orzechowski, Wiesław Bielawski
ZUSAMMENFASSUNG
Storage proteins of cereal seeds are processed during accumulation and degraded during germination primarily by cysteine proteinases. One of the mechanisms controlling the activity of these enzymes is the synthesis of specific inhibitors named phytocystatins. Here we present the complete gene sequence of a triticale ( × Triticosecale Wittm.) phytocystatin, TrcC-6, which encodes a 152-amino acid protein with a putative 25-amino acid signal peptide. This protein has a calculated molecular mass of 16.2 kDa, and was assigned to phylogenetic group B of phytocystatins. Because TrcC-6 transcripts are present in triticale seeds, we hypothesized that this phytocystatin regulates storage protein accumulation and degradation. Therefore, changes in gene expression during the entire period of seed development and germination were examined. TrcC-6 transcripts and TrcC-6 protein levels increased during the maturation of seeds and remained high during the first hours of germination. This enabled us to conclude that TrcC-6 likely regulates seed germination by the regulation of storage protein hydrolysis. For the analysis of TrcC-6 inhibitory activity, recombinant protein was expressed in Escherichia coli BL21 (DE3) and purified. Recombinant TrcC-6 proved to be a potent inhibitor of cysteine proteinases. It inhibited the in vitro activity of papain (EC 3.4.22.2) and ficin (EC 3.4.22.3). Furthermore, native PAGE analysis revealed that recombinant TrcC-6 inhibits the activity of endogenous cysteine proteinases present in germinating seeds of triticale. Based on these results, TrcC-6 is likely one of the important factors that regulate cysteine proteinase activity during the accumulation and mobilization of storage proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Natriumacetat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%