Merck
Biscationic Tartaric Acid-Based Amphiphiles: Charge Location Impacts Antimicrobial Activity.
Langmuir : the ACS journal of surfaces and colloids (2015-10-22)
Allison Faig, Timothy D Arthur, Patrick O Fitzgerald, Michael Chikindas, Evan Mintzer, Kathryn E Uhrich
ZUSAMMENFASSUNG
Cationic amphiphiles have received increasing attention as antimicrobials given their unique ability to disrupt bacteria cell membranes. While extensive research has demonstrated that amphiphiles' hydrophobic-to-charge ratio significantly modulates antibacterial activity, less work has focused on elucidating the specific impact of charge location on amphiphile bioactivity. In this study, two series of cationic amphiphiles, termed bola-like and gemini-like, were synthesized with analogous hydrophobic-to-charge ratios yet differing charge location, and their resulting antibacterial activity was assessed. Bola-like amphiphiles exhibited preferential activity against two Gram-positive bacteria, with activity increasing with increasing hydrophobicity, whereas gemini-like amphiphiles were active against both Gram-positive and Gram-negative bacteria, with activity decreasing with increasing hydrophobicity. After identifying lead compounds from each amphiphile series (bola- and gemini-like), biophysical experiments indicated that both amphiphiles were membrane-active; notably, the lead gemini-like amphiphile exhibited a strong dependence on electrostatic interactions for membrane interaction. In contrast, the lead bola-like amphiphile exhibited a reliance on both hydrophobic and electrostatic contributions. These results demonstrate that charge location significantly impacts cationic amphiphiles' antibacterial and membrane activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1,2-Dioleoyl-sn-Glycero-3-Phosphocholin, lyophilized powder
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.