Merck
Dissipation and Removal of the Etofenprox Residue during Processing in Spring Onion.
Journal of agricultural and food chemistry (2015-07-15)
Kyu-Won Hwang, Woo-Suk Bang, Hyeong-Wook Jo, Joon-Kwan Moon
ZUSAMMENFASSUNG
The dissipation and removal of the etofenprox residue was studied in spring onion grown under greenhouse conditions. Samples of spring onion were collected at 0, 1, 2, 4, 6, 8, 10, 12, and 14 days after last application, and removal rates of etofenprox by washing and drying processes were measured. Samples were extracted with acetone and partitioned with dichloromethane. The dichloromethane layer was then concentrated, cleaned up with florisil column chromatography, and analyzed with high-performance liquid chromatography-ultraviolet detector (HPLC-UVD). At the fortification levels of 0.5, 1.0, and 10 mg/kg, recoveries ranged from 92.0 to 107.7%, with a coefficient of variation of 4.3-7.9% (n = 3). The method limit of quantification (MLOQ) was found to be 0.05 mg/kg in spring onion. The half-lives of etofenprox in spring onion were found to be 9.5 and 7.9 days, at the single or double application rates. Removal rates of etofenprox were 21.6-43.9 and 66.6-88.5% by various washing or drying processes, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.