Merck
  • Carbon Monoxide Releasing Molecule Accelerates Reendothelialization after Carotid Artery Balloon Injury in Rat.
Carbon Monoxide Releasing Molecule Accelerates Reendothelialization after Carotid Artery Balloon Injury in Rat.
Biomedical and environmental sciences : BES (2015-05-15)
Qing Song Hu, Yang Xin Chen, Qing Sheng Huang, Bing Qing Deng, Shuang Lun Xie, Jing Feng Wang, Ru Qiong Nie
ZUSAMMENFASSUNG
This study was aimed to investigate the effects of carbon monoxide releasing molecule (CORM-2), a novel carbon monoxide carrier, on the reendothelialization of carotid artery in rat endothelial denudation model. Male rats subjected to carotid artery balloon injury were treated with CORM-2, inactive CORM-2 (iCORM-2) or dimethyl sulfoxide (DMSO). The reendothelialization capacity was evaluated by Evans Blue dye and the immunostaining with anti-CD31 antibody. The number of circulating endothelial progenitor cells (EPCs) was detected by flow cytometry. The proliferation, migration, and adhesion of human umbilical vein endothelial cells (HUVECs) were assessed by using [3H]thymidine, Boyden chamber and human fibronectin respectively. The expressions of protein were detected by using western blot analysis. CORM-2 remarkably accelerated the re-endothelialization 5 d later and inhibited neointima formation 28 d later. In addition, the number of peripheral EPCs significantly increased in CORM-2-treated rats than that in iCORM-2 or DMSO-treated rats after 5 d later. In vitro experiments, CORM-2 significantly enhanced the proliferation, migration and adhesion of HUVECs. The levels of Akt, eNOS phosphorylation, and NO generation in HUVECs were also much higher in CORM-2 treated group. Blocking of PI3K/Akt/eNOS signaling pathway markedly suppressed the enhanced migration and adhesion of HUVECs induced by CORM-2. CORM-2 could promote endothelial repair, and inhibit neointima formation after carotid artery balloon injury, which might be associated with the function changes of HUVECs regulated by PI3K/Akt/eNOS pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Thymidin, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Thymidin, ≥99.0% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®