Merck
Decellularized porcine aortic intima-media as a potential cardiovascular biomaterial.
Interactive cardiovascular and thoracic surgery (2015-05-15)
Pingli Wu, Naoko Nakamura, Tsuyoshi Kimura, Kwangwoo Nam, Toshiya Fujisato, Seiichi Funamoto, Tetsuya Higami, Akio Kishida
ZUSAMMENFASSUNG
The aim of this research is to investigate the histological and mechanical properties of decellularized aortic intima-media, a promising cardiovascular biomaterial. Porcine aortic intima-media was decellularized using two methods: high hydrostatic pressurization (HHP) and sodium dodecyl sulphate (SDS). The histological properties were characterized using haematoxylin and eosin staining and Elastica van Gieson staining. The mechanical properties were evaluated using a tensile strength test. The structure of the HHP-treated samples was unchanged histologically, whereas that of the SDS-treated samples appeared structurally loose. Consequently, with regard to the mechanical properties of SDS-decellularized intima-media, elastic modulus and tensile strength were significantly decreased. The decellularization method affected the structure and the mechanical properties of the biomaterial. The HHP-treated sample was structurally and mechanically similar to the untreated control. Its mechanical properties were similar to those of human heart valves and the iliac artery and vein. Our results imply that porcine aortic intima-media that is decellularized with HHP is a potential cardiovascular biomaterial.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Trizma® Base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%