Merck
  • Identification and analysis of exosomes secreted from macrophages extracted by different methods.
Identification and analysis of exosomes secreted from macrophages extracted by different methods.
International journal of clinical and experimental pathology (2015-08-12)
Jianjun Wang, Yongliang Yao, Jianhong Wu, Guangxin Li
ZUSAMMENFASSUNG
Exosomes were small vesicles secreted by many cells, and they can play an important role in cell signal transductions. Because the diameter of exosomes is about 30-100 nm, it is so difficult to collection them. In this paper, three kinds of exosomes purifying methods (density gradient ultracentrifugation method, the ultracentrifugation and ultrafiltration method, ExoQuick™ Extraction kit method) were used to collected exosomes in culture supernatants of macrophages. The morphologies of three kinds of exosomes were analyzed by transmission electron microscopy (TEM), and the characteristic molecules such as CD86, LAMP-1, HSP-70 on exosomes were analyzed with Western blot. In addition, the biological activities of exosomes purified by three kinds of methods in vitro were analyzed by ELISA and flow cytometry methods. All experimental results show that the purity and quality of exosomes collected by ExoQuick™ extraction kit and ultracentrifugation and ultrafiltration method were better, and they could enhance the expression of MHC-I in macrophages and promote cells to secrete more TNF-α to cause inflammatory response of macrophages. The analysis pointed out that the advantages and disadvantages of the three methods by biological activities or components of exosomes. Therefore, the ultracentrifugation and ultrafiltration method or the ExoQuick™ Extraction kit method were more suitable to be applied in the scientific research.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®