Merck
The targeting of human and mouse B lymphocytes by dasatinib.
Experimental hematology (2015-02-03)
Morten P Oksvold, Johanna M Duyvestyn, Samantha A Dagger, Samuel J Taylor, Lise Forfang, June H Myklebust, Erlend B Smeland, Wallace Y Langdon
ZUSAMMENFASSUNG
Dasatinib inhibits B-cell receptor-Abelson murine leukemia viral oncogene homologue 1, Src, and other tyrosine kinases. Few studies have addressed the impact of dasatinib on normal blood cells, especially in vivo. Here we show that dasatinib leads to a reduced number of human CD19+ peripheral B cells owing to a strong induction of apoptosis. In contrast, no similar effect on T-cell viability was observed. However, dasatinib induced a comparable broad inhibition of the early events of B- and T-cell receptor signaling. Furthermore, dasatinib was shown to be a more pronounced inhibitor of both basal and B-cell receptor-induced activity of Bruton's tyrosine kinase and PLCγ2 compared with the more specific Bruton's tyrosine kinase inhibitor ibrutinib. Human progenitor B cells from the pre-B stage were sensitive to dasatinib. In an in vivo murine model, dasatinib reduced B-lineage cells in the bone marrow with a marked effect on the pre-B subpopulation. Dasatinib led to a reduced spleen size, with a loss of large immature transitional immunoglobulin M(+)/immunoglobulin D(-) B cells and a reduction in germinal center B cells. Dasatinib caused a marked loss of thymocytes without affecting myeloid lineage cells or hematopoietic progenitors. This study reveals important side effects of dasatinib with specific loss of activated B and thymocyte populations, which may have an impact during long-term treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Phenylessigsäure, 99%
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Phenylessigsäure, ≥99%, FCC, FG
Sigma-Aldrich
DL-Tyrosin, 99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®