Merck
  • Simultaneous determination of valsartan, amlodipine besylate and hydrochlorothiazide using capillary zone electrophoresis (CZE).
Simultaneous determination of valsartan, amlodipine besylate and hydrochlorothiazide using capillary zone electrophoresis (CZE).
Die Pharmazie (2015-07-21)
W Ebeid, M Salim, E Elkady, A Elzahr, R El-Bagary, G Patonay
ZUSAMMENFASSUNG
A capillary zone electrophoresis method was developed for the simultaneous determination of valsartan (VAL), amlodipine besylate (AML) and hydrochlorothiazide (HCZ) in their combined tablets. Separation was achieved on a fused silica capillary by applying a potential of 15 kV (positive polarity) and a running background electrolyte containing 40 mM phosphate buffer at pH 7.5 with UV detection at 230 nm. The samples were injected hydrodynamically for 3s at 0.5 psi and the temperature of the capillary cartridge was kept at 25 degrees C. Pyrazinoic acid was used as an internal standard. The method was validated according to ICH guidelines regarding specificity, linearity, limits of detection and quantitation, accuracy and precision, (Supplementary materials, Table S2). The method showed satisfactory linearity in the ranges of 10-200, 2-20 and 2-20 μg mL(-1) with LODs of 1.82, 0.39, 0.65 μg mL(-1) and LOQs of 5.51, 1.17, 1.96 μg mL(-1) for VAL, AML and HCZ, respectively. The proposed method was successfully applied for the analysis of the studied drugs in their laboratory prepared mixtures and co-formulated tablets. The results were compared with reported methods and no significant differences were found. The proposed method can be used for quality control of the cited drugs in ordinary laboratories.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Natriumphosphat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Natriumphosphat monobasisch -Lösung, BioUltra, 5 M in H2O
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumphosphat, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Pyrazincarbonsäure, 99%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Hydrochlorthiazid, crystalline
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Hydrochlorthiazid, meets USP testing specifications
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Natriumphosphat monobasisch-16O4, 99.9 atom % 16O
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O