Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
ORBO™ Carboxen-564 Röhrchen, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO™ 78 HBr auf Carboxen® 564 speziell gereinigt (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carboxen®-1010 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sigma-Aldrich
Stickstoff, ≥99.998%
Supelco
Carboxen® Adsorptionsmittel, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Sigma-Aldrich
2-Butanon, FCC, FG
Sigma-Aldrich
Palmitoylethanolamide
Supelco
Carboxen®-1006 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
FLM Carbopack™ X TD-Röhrchen aus deaktiviertem Edelstahl, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carboxen®-1010 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carbopack™ Adsorbens, matrix Carbopack™ B, 60-80 mesh, bottle of 10 g
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
Carbopack™ Adsorbens, matrix Carbopack™ C, 80-100 mesh, bottle of 10 g
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Sigma-Aldrich
4-Methyl-2-pentanon, ≥99%, FCC
Supelco
ORBO™-93 Sorbensröhrchen, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
Carbopack™ Adsorbens, matrix Carbopack™ C, 60-80 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Kapillar-GC-Säule, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
SPME-Faseranordnung Carboxen/Polydimethylsiloxan (CAR/PDMS), 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub
Supelco
Carbopack™ Adsorbens, matrix Carbopack™ X, 40-60 mesh, bottle of 10 g