Merck
  • The synergistic killing of AML cells co-cultured with HS-5 bone marrow stromal cells by As2O3 and the PI3K/Akt signaling pathway inhibitor LY294002.
The synergistic killing of AML cells co-cultured with HS-5 bone marrow stromal cells by As2O3 and the PI3K/Akt signaling pathway inhibitor LY294002.
Die Pharmazie (2015-06-13)
Ping Chen, Juanying Wu, Qin Yuan, Xi Jiang, Huifang Huang
ZUSAMMENFASSUNG
We aimed to investigate whether a combination of resistance to arsenic trioxide (As2O3) and the phosphoinositide 3-kinase (PI3K)/Akt signaling pathway inhibitor LY294002 can inhibit the proliferation of AML cells in the bone marrow microenvironment. Three AML cell lines were grown with HS-5 human bone marrow stromal cells in adherent co-cultures. The inhibitory effects of As2O3 alone or in combination with LY294002 on the proliferation of these co-cultured AML cells were observed. The PI3K/Akt signaling pathway was detected by Western Blot in co-cultured AML cells cultured alone or treated with As2O3 alone or in combination with LY294002. Our results demonstrate that AML cells adhered to stroma exhibited significantly reduced sensitivity to As2O3. The resistance can be partially abolished by inhibiting the PI3K/Akt pathway. The administration of As2O3 in combination with a PI3K/Akt signaling pathway inhibitor may be expected to become a new approach to the treatment of AML.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
5-Brom-2′-deoxyuridin, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
5-Brom-2′-deoxyuridin, BioUltra, ≥99%
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®