- Synthesis, Characterization and Thermal Decomposition Kinetics of a Novel Benzofuran Ketoxime Derived Polymer.
Synthesis, Characterization and Thermal Decomposition Kinetics of a Novel Benzofuran Ketoxime Derived Polymer.
Acta chimica Slovenica (2015-06-19)
Zehra Bolat, Cumhur Kırılmış, Adnan Kurt
PMID26085427
ZUSAMMENFASSUNG
A novel benzofuran ketoxime derived polymer, poly(benzofuran-2-yl-methylketoxime-O-methacrylate) [poly(BMKMA], was firstly synthesized by free radical polymerization method. Its thermal degradation studies were performed by thermogravimetric analysis (TGA) in order to determine the actual reaction mechanisms of the decomposition process. The activation energy of the solid-state process was determined using Flynn-Wall-Ozawa method, which resulted to be 235.94 kJ/mol. The activation energies of different mechanism models were determined by Coats-Redfern, Madhusudanan and Van Krevelen kinetic methods. Compared with the Ozawa method, the actual reaction mechanism obeyed deceleration type, phase boundary controlled reaction (R1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture