Merck
  • Beneficial effects of levobupivacaine regional anaesthesia on postoperative opioid induced hyperalgesia in diabetic mice.
Beneficial effects of levobupivacaine regional anaesthesia on postoperative opioid induced hyperalgesia in diabetic mice.
Journal of translational medicine (2015-07-03)
Anne Gomez-Brouchet, Nelly Blaes, Lionel Mouledous, Olivier Fourcade, Ivan Tack, Bernard Francès, Jean-Pierre Girolami, Vincent Minville
ZUSAMMENFASSUNG
Diabetic neuropathy is one of the most common complications of diabetes and causes various problems in daily life. The aim of this study was to assess the effect of regional anaesthesia on post surgery opioid induced hyperalgesia in diabetic and non-diabetic mice. Diabetic and non-diabetic mice underwent plantar surgery. Levobupivacaine and sufentanil were used before surgery, for sciatic nerve block (regional anaesthesia) and analgesia, respectively. Diabetic and non-diabetic groups were each randomly assigned to three subgroups: control, no sufentanil and no levobupivacaine; sufentanil and no levobupivacaine; sufentanil and levobupivacaine. Three tests were used to assess pain behaviour: mechanical nociception; thermal nociception and guarding behaviours using a pain scale. Sufentanil, alone or in combination with levobupivacaine, produced antinociceptive effects shortly after administration. Subsequently, sufentanil induced hyperalgesia in diabetic and non-diabetic mice. Opioid-induced hyperalgesia was enhanced in diabetic mice. Levobupivacaine associated to sufentanil completely prevented hyperalgesia in both groups of mice. The results suggest that regional anaesthesia can decrease opioid-induced hyperalgesia in diabetic as well as in non-diabetic mice. These observations may be clinically relevant for the management of diabetic patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis