Merck
  • CD4⁺ T cells play a crucial role for lenalidomide in vivo anti-tumor activity in murine multiple myeloma.
CD4⁺ T cells play a crucial role for lenalidomide in vivo anti-tumor activity in murine multiple myeloma.
Oncotarget (2015-10-09)
Liang Zhang, Enguang Bi, Sungyoul Hong, Jianfei Qian, Chengyun Zheng, Michael Wang, Qing Yi
ZUSAMMENFASSUNG
Lenalidomide modulates the host immune response against myeloma via multiple actions. Although these effects have been elucidated in vitro, the central action of lenalidomide-mediated anti-myeloma immune response in vivo is not clear. To investigate its immune action in vivo, we selected the murine myeloma cell line 5TGM1, which is resistant to direct tumoricidal effects of lenalidomide in vitro and in immunodeficient mice, but sensitive to lenalidomide treatment in 5TGM1-bearing immunocompetent mice. Depletion of CD4+ T cells, but not NK cells, B cells, or CD8+ T cells, deprived lenalidomide of its therapeutic effects on 5TGM1-bearing immunocompetent mice. Lenalidomide significantly increased the numbers of IFN-γ-secreting CD4+ and CD8+ T cells but had no effects on NK cells and B cells in this mouse model. Lenalidomide slightly decreased the number of CD25+Foxp3+ T cells but increased perforin expression in CD8+ T cells in vivo. Using this mouse model for investigation of anti-tumor immune action of lenalidomide, we demonstrated that lenalidomide facilitated a type-1 anti-tumor immune response in vivo. The CD4+ T cell subset may play a critical role in the lenalidomide-mediated anti-myeloma immune response in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®