Merck
  • A preliminary report of successful cleavage after calcium ionophore activation at ICSI in cases with previous arrest at the pronuclear stage.
A preliminary report of successful cleavage after calcium ionophore activation at ICSI in cases with previous arrest at the pronuclear stage.
Reproductive biomedicine online (2015-10-29)
Ehab Darwish, Yasmin Magdi
ZUSAMMENFASSUNG
Artificial oocyte activation (AOA) has been previously suggested as a means to overcome the problem of total fertilization failure, which affects about 1-3% of the intracytoplasmic sperm injection (ICSI) cycles. A preliminary study on the application of chemical AOA was conducted using A23187 Ca(2+) ionophore to improve embryonic development in four women with a history of complete fertilization arrest and inability to transit to cleavage stage during previous ICSI trials. Data indicated that activated oocytes resulted in better fertilization, embryonic development and clinical pregnancy in one of the four couples. Therefore, ICSI combined with AOA using Ca(2+) ionophore may be useful in selected patients with cleavage failure, and may help the zygotes to reach more advanced developmental stages.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Luteinisierendes Hormon Releasing-Hormon (LHRH), ≥97% (HPLC)
Sigma-Aldrich
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)